About this item
Rugged, compact hotplates with an ergonomic design are ideal for educational labs and can heat up to 1000 mL of water.
- Rugged 5x5" aluminum top plate
- Boils 300 mL of water in 16 minutes
- Built-in support rod holder
- Five-Year Warranty on Parts and Labor
Durable 5x5" cast aluminum top plate will not chip or crack, and provides a uniform heating surface
The fixed temperature hotplate features an illuminated rocker switch to activate the preset fixed temperature of 375°C.
The built-in support rod holder with locking knob accepts an optional support rod and clamp kit. Kit includes 45.7 cm (18”) support rod, thermometer extension clamp, three-prong medium swivel clamp, and hook connector. Optional support plate allows hotplate to be mounted to lab frames or ring stands.
Bi-metallic thermostat offers reliable temperature control. Powerful heater reaches maximum temperature in minutes.
Specifications
- Catalog No:
- 10153-312
- 10153-300
- Overall dimensions:
- 20L x 15.2W x 12.4H cm (7.9 x 6 x 4.9")
- 20L x 15.2W x 12.4H cm (7.9 x 6 x 4.9")
- Shipping weight:
- 1.8 kg (4 lbs.)
- 1.8 kg (4 lbs.)