Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MT3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12179-1-AP
MT3(Metallothionein-3) is also names as GIF(Growth inhibitory factor),GIFB inhibits N-glycosylation of IgE-binding factors and that unglycosylated IgE-binding factors selectively suppressed IgE sybthesis.The major cell source of GIF is antigen-specific suppressor T(Ts)cell.The molecular size of GIF from T cells is 15 kDa, while GIF from macrophages consisted of 40-and 15-kDa molecules.

IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: MT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 12179-1-AP 10090-526 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MT3
Antigen name Metallothionein 3
Reactivity Mouse, Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms GIFB, GIF, GRIF, ZnMT3
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage buffer PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty