|
![]() |
ORDER
SPECIFICATIONS
Antigen symbol | PEPD |
Antigen name | Peptidase D |
Reactivity | Mouse, Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage buffer | PBS with 0.1% sodium azide and 50% glycerol pH 7.3. |
Shipping temperature | Store at -20°C |