![]() |
ORDER
SPECIFICATIONS
Antigen symbol | MAGEA1 |
Antigen name | Melanoma-associated antigen 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Antigen synonyms | melanoma antigen MAGE-1, melanoma antigen family A 1, antigen MZ2-E, cancer/testis antigen 1.1, MAGE-1 antigen, member 1, melanoma-associated antigen MZ2-E, cancer/testis antigen family 1, melanoma-associated antigen 1 |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage buffer | PBS with 0.1% sodium azide and 50% glycerol pH 7.3. |
Shipping temperature | Store at -20°C |