Order Entry
ContactUsLinkComponent
Anti-MCM10 Rabbit Polyclonal Antibody
Anti-MCM10 Rabbit Polyclonal Antibody
Catalog #: 10090-040
Supplier:  Proteintech
Anti-MCM10 Rabbit Polyclonal Antibody
Catalog #: 10090-040
Supplier:  Proteintech
Price
$726.33
Log in for availability
Unit of Measure Each (150µl)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Minichromosome Maintenance Complex Component 10
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    MCM10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Western blot:
    Yes
  • Antigen synonyms:
    PRO2249,
    DNA43,
    CNA43
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10090-040

Specifications

About this item

MCM10, also named as HsMCM10, belongs to the MCM10 family. It acts as a replication initiation factor that brings together the MCM2-7 helicase and the DNA polymerase alpha/primase complex in order to initiate DNA replication. Additionally, it plays a role in preventing DNA damage during replication and is essential for the integrity of the RECQ4–MCM replicative helicase complex. It mediates RECQ4 association with MCM2-7 helicase complex during DNA replication.

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: MCM10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought