![]() |
ORDER
SPECIFICATIONS
Antigen symbol | ETS2 |
Antigen name | V-Ets Avian Erythroblastosis Virus E2 Oncogene Homolog 2 |
Reactivity | Mouse, Human, Rat |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
ImmunoPrecipitation | Yes |
Antigen synonyms | Oncogene ETS2, ETS2, Protein C-ets-2, ETS2_HUMAN, V ets erythroblastosis virus E26 oncogene homolog 2., C ets 2 protein |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage buffer | PBS with 0.1% sodium azide and 50% glycerol pH 7.3. |
Shipping temperature | Store at -20°C |