Anti-NCR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
55118-1-AP
10084-334EA 635.16 CAD
10084-334
Anti-NCR2 Rabbit Polyclonal Antibody
Antibodies
CD336, also named as NCR2, LY95 and NKp44, belongs to the natural cytotoxicity receptor (NCR) family. It is cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis. It is expressed on activated human NK cells. CD336 displays a single extracellular Ig-like V domain and a transmembrane portion containing the charged residue (Lysine), likely involved in the association with KARAP/DAP12 molecules. The antibody is specific to CD336.

Western Blot: Human spleen Tissue, 1:200-1:2000

Type: Primary
Antigen: NCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR