Anti-PPP3CB Rabbit Polyclonal Antibody

Supplier: Proteintech
55148-1-AP
10081-402EA 635.16 CAD
10081-402
Anti-PPP3CB Rabbit Polyclonal Antibody
Antibodies
PPP3CB, also named as CALNA2, CALNB and CNA2, belongs to the PPP phosphatase family and PP-2B subfamily. It is a calcium-dependent, calmodulin-stimulated protein phosphatase. It may have a role in the calmodulin activation of calcineurin. This antibody is special to PPP3CB.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: PPP3CB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR