Anti-PARP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
55149-1-AP
10081-706EA 635.16 CAD
10081-706
Anti-PARP2 Rabbit Polyclonal Antibody
Antibodies
PARP2, also named as ADPRT2 and ADPRTL2, is involved in the base excision repair (BER) pathway, by catalyzing the poly(ADP-ribosyl)ation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signaling pathway leading to the reparation of DNA strand breaks. This antibody is specific to PARP2 (N-terminal).

Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: PARP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR