Anti-THOC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
55178-1-AP
10095-946EA 635.16 CAD
10095-946
Anti-THOC2 Rabbit Polyclonal Antibody
Antibodies
THOC2 also termed as THO complex 2 or CXorf3 is 1593 amino acid protein, which belongs to the THOC2 family. THOC2 is a component of heteromultimeric THO/TREX (transcription/export) complex, which is a multi-protein structure consisting of THOC1, THOC2, THOC3, BAT1 and ALY. The TREX complex associates with spliced mRNA and not with unspliced pre-mRNA. It is recruited to spliced mRNAs by a transcription-independent mechanism.

IP: HeLa Cells, 1:200-1:1000; Western Blot: HeLa Cells, 1:200-1:1000; IF: HeLa Cells, 1:20-1:200

Type: Primary
Antigen: THOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR