Anti-L-Plastin Rabbit Polyclonal Antibody

Supplier: Proteintech
55208-1-AP
10081-280EA 635.16 CAD
10081-280
Anti-L-Plastin Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:1000-1:10000

Type: Primary
Antigen: L-Plastin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR