Anti-HCN4 Rabbit Polyclonal Antibody

Supplier: Proteintech
55224-1-AP
10088-110EA 635.16 CAD
10088-110
Anti-HCN4 Rabbit Polyclonal Antibody
Antibodies
HCN4 belongs to the potassium channel HCN family. It hyperpolarization-activated ion channel with very slow activation and inactivation exhibiting weak selectivity for potassium over sodium ions. HCN4 may contribute to the native pacemaker currents in heart (If) and in neurons (Ih). It is activated by cAMP. HCN4 may mediate responses to sour stimuli. . Defects in HCN4 are a cause of sick sinus syndrome type 2 (SSS2). Defects in HCN4 are the cause of Brugada syndrome type 8 (BRS8). This antibody is specific to HCN4.

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: HCN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR