Anti-ESR2 beta-6 Rabbit Polyclonal Antibody

Supplier: Proteintech
55471-1-AP
10086-658EA 635.16 CAD
10086-658
Anti-ESR2 beta-6 Rabbit Polyclonal Antibody
Antibodies
Estrogen receptor-beta (ESR2) is a member of the superfamily of nuclear receptors, which can transduce extracellular signals into transcriptional responses. It binds estrogens with an affinity similar to that of ESR1, and activates expression of reporter genes containing estrogen response elements (ERE) in an estrogen-dependent manner. DNA-binding by ESR1 and ESR2 is rapidly lost at 37 degrees Celsius in the absence of ligand while in the presence of 17 beta-estradiol and 4-hydroxy-tamoxifen loss in DNA-binding at elevated temperature is more gradual

IHC: Human cervix, 1:20-1:200

Type: Primary
Antigen: ESR2 beta-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR