Anti-HOXA11 Rabbit Polyclonal Antibody

Supplier: Proteintech
55495-1-AP
10088-410EA 635.16 CAD
10088-410
Anti-HOXA11 Rabbit Polyclonal Antibody
Antibodies
HOXA11 belongs to the Abd-B homeobox family. It is a sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Defects in HOXA11 are the cause of radioulnar synostosis with amegakaryocytic thrombocytopenia (RSAT). This antibody is specific to Human/Mouse HOXA11 and Rat Hoxa10.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: HOXA11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR