Anti-STX6 Mouse Monoclonal Antibody [clone: 2B6G4]

Supplier: Proteintech
60059-1-IG
10095-534EA 635.16 CAD
10095-534
Anti-STX6 Mouse Monoclonal Antibody [clone: 2B6G4]
Antibodies
Syntaxin-6 (STX6) is a SNARE protein located in the trans-Golgi network (TGN) and endosomes. It associates with a variety of SNARE proteins and is involved in intracellular vesicle trafficking. STX6 is required for sorting proteins from endosomes toward either the TGN or lysosomes.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: STX6
Clonality: Monoclonal
Clone: 2B6G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR