Anti-BRD8 Mouse Monoclonal Antibody [clone: 5B6D6]

Supplier: Proteintech
60121-1-IG
10083-672EA 635.16 CAD
10083-672
Anti-BRD8 Mouse Monoclonal Antibody [clone: 5B6D6]
Antibodies
BRD8 is one of the bromodomain-containing proteins, which is an acetylated lysine- binding domain and thought to be involved in regulation of protein acetylation and/or HAT (histone acetyl transferase) activity . It is also reported to be an accessory subunit of NuA4 HAT complex (also known as TRRAP/TIP60 complex) through biochemical purification . In addition, it can bind to thyroid hormone receptor-ß and/or retinoid X receptor, and can act as a coactivator of the nuclear hormone receptor-mediated transcription in reporter assays .

Western Blot:fetal Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: BRD8
Clonality: Monoclonal
Clone: 5B6D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR