Anti-RABEP2 Mouse Monoclonal Antibody [clone: 1A4A4]

Supplier: Proteintech
60125-1-IG
10093-242EA 635.16 CAD
10093-242
Anti-RABEP2 Mouse Monoclonal Antibody [clone: 1A4A4]
Antibodies
RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.

Western Blot:PC-3 Cells, 1:200-1:1000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RABEP2
Clonality: Monoclonal
Clone: 1A4A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR