Anti-NENF Mouse Monoclonal Antibody [clone: 4G9E12]

Supplier: Proteintech
60131-1-IG
10091-034EA 635.16 CAD
10091-034
Anti-NENF Mouse Monoclonal Antibody [clone: 4G9E12]
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human cerebellum Tissue, 1:20-1:200

Type: Primary
Antigen: NENF
Clonality: Monoclonal
Clone: 4G9E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR