Anti-GSTO1 Mouse Monoclonal Antibody [clone: 5E1E10]

Supplier: Proteintech
60150-2-IG
10087-934EA 635.16 CAD
10087-934
Anti-GSTO1 Mouse Monoclonal Antibody [clone: 5E1E10]
Antibodies
Western Blot:Jurkat Cells, 1:200-1:1000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: GSTO1
Clonality: Monoclonal
Clone: 5E1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR