Anti-WTAP Mouse Monoclonal Antibody [clone: 4A10G9]

Supplier: Proteintech
60188-1-IG
10097-152EA 849.57 CAD
10097-152
Anti-WTAP Mouse Monoclonal Antibody [clone: 4A10G9]
Antibodies
WTAP, also named as KIAA0105 and hFL(2)D, is previously identified as a protein associated with Wilms' tumor-1 (WT-1) protein that is essential for the development of the genitourinary system. It is a Pre-mRNA-splicing regulator protein. It is a regulatory subunit of the WMM N6-methyltransferase complex which plays a role in the efficiency of mRNA splicing and processing and mRNA stability. It is required for accumulation of METTL3 and METTL14 to nuclear speckle.

Western Blot: Hepg2 Cells, 1:500-1:5000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: WTAP
Clonality: Monoclonal
Clone: 4A10G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR