Anti-CD23 Mouse Monoclonal Antibody [clone: 4C7C9]

Supplier: Proteintech
60208-2-IG
10082-008EA 635.16 CAD
10082-008
Anti-CD23 Mouse Monoclonal Antibody [clone: 4C7C9]
Antibodies
CD23, also known as low affinity immunoglobulin epsilon Fc receptor, is a transmembrane glycoprotein present on a subpopulation of B lymphocytes in germinal centres, EBV-transformed B-lymphoblastoid cell lines, follicular dendritic cells, and a subpopulation of peripheral blood cells. CD23 has essential roles in the regulation of IgE production and in the differentiation of B-cells.

Western Blot: Hela Cells, 1:500-1:5000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: 4C7C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR