Anti-COXIV Mouse Monoclonal Antibody [clone: 3C7D2]

Supplier: Proteintech
60251-1-IG
10082-038EA 569.01 CAD
10082-038
Anti-COXIV Mouse Monoclonal Antibody [clone: 3C7D2]
Antibodies
COX4I1, also named as COX4 and COXIV-1, belongs to the cytochrome c oxidase IV family. It is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport. COX4I1 is a marker for mitochondria. It has two isoforms (isoform 1 and 2). Isoform 1(COX4I1) is ubiquitously expressed and isoform 2 is highly expressed in lung tissues. COX4I1 is commonly used as a loading control. This antibody was generated against full length COX4I1 protein and cross reacts with COX4I2.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: HeLa Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: COXIV
Clonality: Monoclonal
Clone: 3C7D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR