Anti-ANXA4 Mouse Monoclonal Antibody [clone: 1A1B5]

Supplier: Proteintech
60288-2-IG
10082-926EA 635.16 CAD
10082-926
Anti-ANXA4 Mouse Monoclonal Antibody [clone: 1A1B5]
Antibodies
Annexin IV /placental anticoagulant protein II (ANX4) is a member of the annexin or lipocortin family of calcium-dependent phospholipid binding proteins, and is located at 2p13. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 shares 45 to 59% identity with other members and possibly interacts with ATP. ANX4 has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: ANXA4
Clonality: Monoclonal
Clone: 1A1B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR