Anti-EXOC2 Mouse Monoclonal Antibody [clone: 1C11G2]

Supplier: Proteintech
66011-1-IG
10094-256EA 635.16 CAD
10094-256
Anti-EXOC2 Mouse Monoclonal Antibody [clone: 1C11G2]
Antibodies
EXOC2 (exocyst complex component 2), also known as SEC5 and SEC5L1, is a component of the exocyst complex, and is required to mediate RalB-dependent survival signals in transformed cells. The exocyst complex, composed of eight evolutionarily conserved subunits (SEC3, SEC5, SEC6, SEC8, SEC10, SEC15, EXO70, and EXO84), is involved in tethering post-Golgi secretory vesicles to specific plasma membrane domains. The gene of EXOC2 maps to chromosome 6p25.3, and encodes a 924-amino acid protein with an experimentally determined molecular mass of 95-100 kDa. EXOC2 mRNA is widely expressed with highest levels in brain and placenta.

Western Blot: Human Brain, 1:500-1:5000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: EXOC2
Clonality: Monoclonal
Clone: 1C11G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR