Anti-TPP2 Mouse Monoclonal Antibody [clone: 5H6F11]

Supplier: Proteintech
66017-1-IG
10096-234EA 635.16 CAD
10096-234
Anti-TPP2 Mouse Monoclonal Antibody [clone: 5H6F11]
Antibodies
TPPII(Tripeptidyl-peptidase 2) is an evolutionary conserved serine peptidase of the subtilisin family that removes tripeptides from the free NH2 terminus of oligopeptides. The 138-kDa TPP2 polypeptide forms large oligomeric complexes of >1,000 kDa that are detected in the cytoplasm of all cell types. In addition, a membrane associated TPP 2 isoform is present in most cells.

Western Blot: Hela Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: TPP2
Clonality: Monoclonal
Clone: 5H6F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR