Anti-AHCY Mouse Monoclonal Antibody [clone: 1F9B1]

Supplier: Proteintech
66019-1-IG
10082-620EA 635.16 CAD
10082-620
Anti-AHCY Mouse Monoclonal Antibody [clone: 1F9B1]
Antibodies
AHCY, also named as SAHH, belongs to the adenosylhomocysteinase family. AHCY is a competitive inhibitor of S-adenosyl-L-methionine-dependent methyl transferase reactions; therefore AHCY may play a key role in the control of methylations via regulation of the intracellular concentration of adenosylhomocysteine. AHCY catalyzes the reversible hydrolysis of S-adenosylhomocysteine to adenosine and L-homocysteine..

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: AHCY
Clonality: Monoclonal
Clone: 1F9B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR