Anti-ALB Mouse Monoclonal Antibody [clone: 4A1C11]

Supplier: Proteintech
66051-1-IG
10082-748EA 766.96 CAD
10082-748
Anti-ALB Mouse Monoclonal Antibody [clone: 4A1C11]
Antibodies
Albumin is the most abundant protein in blood plasma. Alterations of level of serum albumin are linked to variety of diseases. Albumin is expressed exclusively by well-differentiated hepatocytes, thus anti-albumin has been used to mark hepatocytes. (21388516, 23832071)

Western Blot: Human Liver, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 4A1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR