Anti-Antithrombin III Mouse Monoclonal Antibody [clone: 2B2E5]

Supplier: Proteintech
66052-1-IG
10093-564EA 635.16 CAD
10093-564
Anti-Antithrombin III Mouse Monoclonal Antibody [clone: 2B2E5]
Antibodies
SERPINC1, also named Antithrombin III or Antithrombin, is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency.

Western Blot:L02 Cells, 1:500-1:5000

Type: Primary
Antigen: Antithrombin III
Clonality: Monoclonal
Clone: 2B2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR