Anti-ARHGDIB Mouse Monoclonal Antibody [clone: 3E7B7]

Supplier: Proteintech
66054-1-IG
10083-086EA 635.16 CAD
10083-086
Anti-ARHGDIB Mouse Monoclonal Antibody [clone: 3E7B7]
Antibodies
ARHGDIB, also known as RhoGDI2, is a conserved member of the RhoGDI family and plays an important role in cell migrations. It regulates the GDP/GTP exchange reaction of the Rho proteins by inhibiting the dissociation of GDP from them, and the subsequent binding of GTP to them. It is mainly in hematopoietic, endothelial, and epithelial cells. It has been linked to tumorigenesis and metastasis. RhoGDI2 expression is downregulated in several cancer types, such as bladder, lung and lymphoma, but is upregulated in prostate and gastric cancer.

Western Blot:U937 Cells, 1:500-1:5000

Type: Primary
Antigen: ARHGDIB
Clonality: Monoclonal
Clone: 3E7B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR