Anti-CPT1C Mouse Monoclonal Antibody [clone: 5D11B5]

Supplier: Proteintech
66072-1-IG
10085-148EA 635.16 CAD
10085-148
Anti-CPT1C Mouse Monoclonal Antibody [clone: 5D11B5]
Antibodies
CPT1C, also named as CATL1, CPTI-B and B-CPTI, belongs to the carnitine/choline acetyltransferase family. Carnitine palmitoyltransferase (CPT) deficiencies are common disorders of mitochondrial fatty acid oxidation. The CPT system is made up of two separate proteins located in the outer (CPT1) and inner (CPT2) mitochondrial membranes. CPT1C is an active forms of related brain-type carnitine palmitoyltransferase I. CPT1C may be a regulated target of malonyl-CoA that relays the “malonyl-CoA signal” in hypothalamic neurons that express the orexigenic and anorexigenic neuropeptides that regulate food intake and peripheral energy expenditure. This antibody can bind the close sequences genes.

Western Blot: Human Brain, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: CPT1C
Clonality: Monoclonal
Clone: 5D11B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR