Anti-TELO2 Mouse Monoclonal Antibody [clone: 3H10F4]

Supplier: Proteintech
66077-1-IG
10095-774EA 635.16 CAD
10095-774
Anti-TELO2 Mouse Monoclonal Antibody [clone: 3H10F4]
Antibodies
Western Blot:A431 Cells, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: TELO2
Clonality: Monoclonal
Clone: 3H10F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR