Anti-ORM1 Mouse Monoclonal Antibody [clone: 5F2H7]

Supplier: Proteintech
66097-1-IG
10091-610EA 635.16 CAD
10091-610
Anti-ORM1 Mouse Monoclonal Antibody [clone: 5F2H7]
Antibodies
Alpha-1-acid glycoprotein 1 (AGP1), also called orosomucoid-1 (ORM1), is a glycoprotein synthesized mostly by hepatocytes and present in human plasma. ORM1 is an acute-phase reactant protein controlled by glucocorticoids, interleukin-1 and interleukin-6, and increase up to 5-50 times upon infection and/or inflammation. Anti-apoptotic effect and role as immunomodulator of ORM have been reported. ORM is an important carrier for synthetic drugs and influences their distribution and availability in the body. This antibody recognizes a band about 44 kDa in human plasma which may be due to the glycosylation of ORM1 or the dimer formation of the protein.

Western Blot: Human Blood, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ORM1
Clonality: Monoclonal
Clone: 5F2H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR