Anti-HBG1 Mouse Monoclonal Antibody [clone: 2A4B5]

Supplier: Proteintech
66168-1-IG
10088-074EA 635.16 CAD
10088-074
Anti-HBG1 Mouse Monoclonal Antibody [clone: 2A4B5]
Antibodies
IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: HBG1-Specific
Clonality: Monoclonal
Clone: 2A4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR