Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Heat-Resistant Brushes
https://ca.vwr.com/stibo/smallweb/std.lang.all/53/14/18945314.jpg
VWR offers heat-resistant brushes appropriate for cleaning a variety of parts and surfaces. Brush handles of stainless steel or durable plastic can withstand autoclaving and be used in cleanrooms. Soft nylon fiber bristles are made for cleaning delicate surfaces and parts, while stronger stainless steel bristles are essential when contamination or rust are present. Some heat-resistant brushes feature mechanical fiber clamping, while others offer epoxy set bristles for deterioration resistance. Select an appropriate bristle trim length and order products individually or in multipacks.
SearchPresentationType-VERTICAL
SearchResultCount:"3"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Gordon Brush
Description:   Cleanroom brushes are durable and non-shedding
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Excelta
Description:   Heat Resistant Brushes have nylon fibers epoxy set into the Delrin handles, to resist deterioration in an autoclave
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Supplier:  Gordon Brush
Description:   Heat-tolerant brushes have stainless steel handles and are durable, allowing them to withstand most sterilizing environments
Product available on GSA Advantage® Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 3  of 3