Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

precision-brushes

https://ca.vwr.com/stibo/smallweb/std.lang.all/96/27/18709627.jpg

VWR offers precision brushes for your lab's specific needs. For cleaning of button samples for assays, select a double head brush with nylon bristles. Choose a frozen section brush with angled boar bristles to prevent curling. Cell cleaning brushes offer firm nylon bristles for easy cell cleaning and removal of solid samples. Instrument and valve cleaning brushes are available. Offered in several sizes, tracheal tube brushes are disposable, and latex free. Purchase precision brushes individually or in multipacks.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (10163-460)
Supplier:  Sklar
Description:   The Valve Cleaning Brush features a double end, with 9mm and 5mm tips. Nonsterile and latex-free.
Supplier:  Sklar
Description:   Disposable tracheal tube brushes are nonsterile and latex-free.
Catalog Number: (17210-008)
Supplier:  Mill-Rose
Description:   Stiff black nylon bristles for quick cleaning button samples for assays.
Supplier:  Gordon Brush
Description:   Brushes are designed for cleaning in tight areas. The brush fibers are mechanically clamped into a stainless steel tube inserted into a Delrin® handle...
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Restek
Description:   Precision cell cleaning brushes produced by Restek are durable and efficient at removing solid samples from cells.
Supplier:  Remco Products
Description:   This pastry brush is supersoft and does not damage the pastry when applying egg, oil, butter, etc.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 6  of 6