Fmoc-Met(O2)-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Met(O₂)-OHN-Fmoc-L-methionine sulphone

H63033-03 H63033-06 H63033-MD
AAH63033-03EA 158.41 CAD
AAH63033-03 AAH63033-06 AAH63033-MD
Fmoc-Met(O2)-OH 98%
Fmoc-Met(O2)-OH
Formula: C₂₀H₂₁NO₆S
Storage Temperature: Ambient
MDL Number: MFCD00153361
CAS Number: 163437-14-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR