1-Benzyl-L-aspartate 98%

Synonyms: L-Aspartic Acid 1-Benzyl EsterH-Asp-OBzl

H63128-06 H63128-03
AAH63128-03EA 68.73 CAD
AAH63128-03 AAH63128-06
1-Benzyl-L-aspartate 98%
1-Benzyl-L-aspartate
Formula: C₁₁H₁₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00063186
CAS Number: 7362-93-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR