1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63627-MD
H63627-06
H63627-03
AAH63627-03EA
285.98
CAD
AAH63627-03
AAH63627-MD
AAH63627-06
1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid 97%
1-[2-(Boc-amino)ethyl]indole-6-carboxylic acid
Formula:
C₁₆H₂₀N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD04973974 CAS Number: 885266-67-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...