5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine 95%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine

H64470-MD H64470-03 H64470-06
AAH64470-03EA 95.43 CAD
AAH64470-03 AAH64470-MD AAH64470-06
5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine 95%
5-Bromo-4-(trifluoromethyl)pyrimidin-2-amine
Formula: C₅H₃BrF₃N₃
Storage Temperature: Ambient
MDL Number: MFCD09261254
CAS Number: 935534-47-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR