5-Bromo-2-(trifluoromethyl)pyrimidine 95%

Supplier: Thermo Scientific Chemicals

Warning

H64899-03 H64899-MD
AAH64899-MDEA 264.82 CAD
AAH64899-MD AAH64899-03
5-Bromo-2-(trifluoromethyl)pyrimidine 95%
5-Bromo-2-(trifluoromethyl)pyrimidine
Formula: C₅H₂BrF₃N₂
MW: 226.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09754046
CAS Number: 799557-86-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR