Cross Connectors, Eldon James
Supplier: Eldon
X0-2NK X0-1NK X0-4NK
89093-296PK 101.25 CAD
89093-296 89093-294 89093-290
Cross Connectors, Eldon James
Connectors Connectors, Tubing
These connectors are made of Natural Kynar® material, which has a temperature range of -62°C to 107°C (-80 to 226°F), and a pressure of 260 psi at 21°C (70°F)
This material can be autoclaved to 140°C (285°F) and meets USP Class VI requirements. Generally, chemically resistant to most acids, salts, weak bases, halogenated solvents, alcohols, nuclear and UV radiation, oxidants, ozone and deionized water.
Certifications: RoHS Compliant.