Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories

Supplier: EPPENDORF BIOTOOLS CA



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description For Use With Supplier No. VWR Catalog Number Unit Price Quantity
Buckets for 4 x 2.0 mL, 2 pcs. Rotor S-24-11-AT of Centrifuges 5430/R and Centrifuge 5427 R 5409721003 75875-166 Each Retrieving
Rotor Lid Rotor F-45-30-11 5409709003 75875-150 Each Retrieving
Rotor Lid Rotor F-45-48-11 5409713000 75875-154 Each Retrieving
Rotor Lid, Aerosol-tight Rotor FA-45-30-11 5409707000 75875-148 Each Retrieving
Rotor Lid, Aerosol-tight Rotor FA-45-24-11-Kit 5409705008 75875-146 Each Retrieving
Rotor Lid, Aerosol-tight Rotor FA-45-12-17 5409701002 75875-142 Each Retrieving
Rotor Lid, Aerosol-tight Rotor FA-45-48-11 5409711008 75875-152 Each Retrieving
Rotor Lid, Aerosol-tight Rotor FA-45-24-11 5409703005 75875-144 Each Retrieving
Rotor Lid, Aerosol-tight Rotor S-24-11-AT 5409720007 75875-164 Each Retrieving
Spare sealing, 5 pcs. FA-45-48-11 Rotor Lid 5409718002 75875-160 Each Retrieving
Spare sealing, 5 pcs. FA-45-12-17 Rotor Lid 5409716000 75875-156 Each Retrieving
Spare sealing, 5 pcs. FA-45-24-11-Kit 5820767006 75875-176 Each Retrieving
Pack of 5 Retrieving
Spare sealing, 5 pcs. S-24-11-AT Rotor Lid 5409719009 75875-162 Each Retrieving
Spare sealing, 5 pcs. FA-45-30-11 Rotor Lid 5820762004 75875-174 Each Retrieving
Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty