Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SJ25C1]

Supplier: Tonbo Biosciences
60-0198-T100 60-0198-T500 60-0198-T025
10208-458EA 2447.25 CAD
10208-458 10208-454 10208-456
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: SJ25C1]
Antibodies
The SJ25C1 antibody reacts with human CD19, a 95 kDa glycoprotein which acts as a co-receptor, along with CD21 (CR2), CD81 (TAPA-1) and CD225 (Leu13), in support of the functional B cell receptor (BCR). This complex provides antigen-specific recognition and subsequent activation of B cells to proliferate and differentiate into antibody-secreting cells (plasma cells) or memory B cells, which are crucial for secondary antigen encounter. Upon activation and tyrosine phosphorylation, the CD19 molecule can provide an anchor for cytoplasmic signaling proteins such as GRB2, SOS or PLCG2. CD19 is a lineage-differentiation marker, as its expression is detectable at the earliest B cell stages, through development, and is finally lost upon transition to mature plasma cells.

The SJ25C1 antibody is widely used as a phenotypic marker for CD19 expression on B cells, as well as on dendritic cell subsets.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR