Anti-LIPI Rabbit Polyclonal Antibody

Supplier: Proteintech
24300-1-AP
10155-164EA 635.16 CAD
10155-164
Anti-LIPI Rabbit Polyclonal Antibody
Antibodies
The LIPI antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LIPI. This antibody recognizes human, mouse antigen. The LIPI antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: LIPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR