Anti-GPATCH2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24366-1-AP-150UL
10155-178EA 635.16 CAD
10155-178
Anti-GPATCH2 Rabbit Polyclonal Antibody
Antibodies
The GPATCH2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human GPATCH2. This antibody recognizes human, mouse antigen. The GPATCH2 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 1000 ; IHC 1 : 50 ; IF 1 : 50

Type: Primary
Antigen: GPATCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR