Anti-PPP1R14D Rabbit Polyclonal Antibody

Supplier: Proteintech
22080-1-AP
10155-024EA 635.16 CAD
10155-024
Anti-PPP1R14D Rabbit Polyclonal Antibody
Antibodies
The PPP1R14D antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PPP1R14D. This antibody recognizes human, mouse antigen. The PPP1R14D antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: PPP1R14D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR