Anti-ACSM5 Rabbit Polyclonal Antibody

Supplier: Proteintech
16591-1-AP
10154-682EA 635.16 CAD
10154-682
Anti-ACSM5 Rabbit Polyclonal Antibody
Antibodies
The ACSM5 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human ACSM5. This Antibody recognizes human,mouse,rat Antigen. The ACSM5 Antibody has been validated for the following applications: ELISA, WB, IHC analysis.

IHC 1 : 50 ; WB 1 : 300

Type: Primary
Antigen: ACSM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR