Anti-BCMO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23308-1-AP
10155-110EA 635.16 CAD
10155-110
Anti-BCMO1 Rabbit Polyclonal Antibody
Antibodies
The BCMO1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human BCMO1. This antibody recognizes human, mouse antigen. The BCMO1 antibody has been validated for the following applications: ELISA, WB, IHC analysis.

IHC 1 : 50

Type: Primary
Antigen: BCMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR