Anti-HOXD13 Rabbit Polyclonal Antibody

Supplier: Proteintech
23520-1-AP
10155-118EA 635.16 CAD
10155-118
Anti-HOXD13 Rabbit Polyclonal Antibody
Antibodies
The HOXD13 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human HOXD13. This antibody recognizes human antigen. The HOXD13 antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; IF 1 : 50

Type: Primary
Antigen: HOXD13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR