Anti-ZER1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16647-1-AP
10154-692EA 635.16 CAD
10154-692
Anti-ZER1 Rabbit Polyclonal Antibody
Antibodies
The ZER1 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human ZER1. This Antibody recognizes human,mouse,rat Antigen. The ZER1 Antibody has been validated for the following applications: ELISA, WB, IP, IHC analysis.

WB 1 : 300

Type: Primary
Antigen: ZER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR